missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3MD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | SH3MD2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SH3MD2 Polyclonal specifically detects SH3MD2 in Human samples. It is validated for Western Blot.Specifications
| SH3MD2 | |
| Polyclonal | |
| Rabbit | |
| Zinc Finger | |
| EC 6.3.2, EC 6.3.2.-, KIAA1494SH3 multiple domains protein 2, Plenty of SH3s, POSHFLJ21602, Protein POSH, putative E3 ubiquitin-protein ligase SH3RF1, RING finger protein 142, RNF142plenty of SH3 domains, SH3 domain containing ring finger 1, SH3 domain-containing RING finger protein 1, SH3 multiple domains 2, SH3MD2 | |
| SH3RF1 | |
| IgG | |
| 93 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q7Z6J0 | |
| 57630 | |
| Synthetic peptides corresponding to SH3RF1(SH3 domain containing ring finger 1) The peptide sequence was selected from the middle region of SH3RF1. Peptide sequence LLKLLSGASTKRKPRVSPPASPTLEVELGSAELPLQGAVGPELPPGGGHG. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title