missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3MD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54972
This item is not returnable.
View return policy
Description
SH3MD2 Polyclonal specifically detects SH3MD2 in Human samples. It is validated for Western Blot.
Specifications
| SH3MD2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 6.3.2, EC 6.3.2.-, KIAA1494SH3 multiple domains protein 2, Plenty of SH3s, POSHFLJ21602, Protein POSH, putative E3 ubiquitin-protein ligase SH3RF1, RING finger protein 142, RNF142plenty of SH3 domains, SH3 domain containing ring finger 1, SH3 domain-containing RING finger protein 1, SH3 multiple domains 2, SH3MD2 | |
| Rabbit | |
| 93 kDa | |
| 100 μL | |
| Zinc Finger | |
| 57630 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q7Z6J0 | |
| SH3RF1 | |
| Synthetic peptides corresponding to SH3RF1(SH3 domain containing ring finger 1) The peptide sequence was selected from the middle region of SH3RF1. Peptide sequence LLKLLSGASTKRKPRVSPPASPTLEVELGSAELPLQGAVGPELPPGGGHG. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction