missing translation for 'onlineSavingsMsg'
Learn More

SGTA Mouse anti-Human, Clone: 1.90E+09, Novus Biologicals™

Mouse Monoclonal Antibody

£234.00 - £378.00

Specifications

Antigen SGTA
Clone 1900000000
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1036, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18242965
View Documents
Novus Biologicals
NBP2-59426-50UL
50 μL
£234.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18243522
View Documents
Novus Biologicals
NBP2-59426-100UL
100 μL
£378.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SGTA Monoclonal specifically detects SGTA in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

SGTA
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Human
alphaSGT, Alpha-SGT, protein containing three tetratricopeptide repeats, SGT1, SGThSGT, small glutamine-rich tetratricopeptide repeat (TPR)-containing, small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha, small glutamine-rich tetratricopeptide repeat-containing protein alpha, UBP, Vpu-binding protein
SGTA
IgG1 κ
Protein A purified
1900000000
Western Blot 1:1000, Flow Cytometry 1:10-1:1036, ELISA, Immunocytochemistry/Immunofluorescence 1:100
Monoclonal
Purified
RUO
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
6449
Recombinant human SGTA (1-313aa) purified from E. coli
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.