missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGTA Mouse anti-Human, Clone: 1.90E+09, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59426-50UL
This item is not returnable.
View return policy
Description
SGTA Monoclonal specifically detects SGTA in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| SGTA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1036, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| alphaSGT, Alpha-SGT, protein containing three tetratricopeptide repeats, SGT1, SGThSGT, small glutamine-rich tetratricopeptide repeat (TPR)-containing, small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha, small glutamine-rich tetratricopeptide repeat-containing protein alpha, UBP, Vpu-binding protein | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6449 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1900000000 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| SGTA | |
| Recombinant human SGTA (1-313aa) purified from E. coli | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction