missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGLT2/SLC5A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £290.00
Specifications
| Antigen | SGLT2/SLC5A2 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388941
|
Novus Biologicals
NBP3-15989-20UL |
20 μg |
£149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18373543
|
Bio-Techne
NBP3-15989-100UL |
100 μg |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SGLT2/SLC5A2 Polyclonal antibody specifically detects SGLT2/SLC5A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| SGLT2/SLC5A2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS, 50% glycerol, pH7.3 | |
| 6524 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 2, SGLT2sodium/glucose cotransporter 2, solute carrier family 5 (sodium/glucose cotransporter), member 2, solute carrier family 5 (sodium/glucose transporter), member 2, Solute carrier family 5 member 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 564-624 of human SGLT2/SLC5A2 (NP_003032.1). RHSKEEREDLDADEQQGSSLPVQNGCPESAMEMNEPQAPAPSLFRQCLLWFCGMSRGGVGS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title