missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ PerkinElmer Split / Splitless Tapered FocusLiner

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan Scientific and Medical™ 09209525

Code : 13

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12944031

  • £730.00 / Pack of 25
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with PerkinElmer instruments
Specifications

Specifications

GC Inlet Liner
92 mm
6.2 mm
25/Pk.
4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers