Learn More
SGE™ SilTite™ FingerTite Ferrule System Starter Kits
For gas chromatography systems
Brand: SGE™ 073615
Code : NEW
Additional Details : Weight : 0.01000kg
Description
SilTite FingerTite is the next generation ferrule system for gas chromatography systems delivering an easy, leak free installation for capillary columns without the use of any tools. Each starter kit includes all the parts necessary to convert one GC system (one injector and one detector) to the SilTite FingerTite system. The system requires no permanent instrument modifications—switching back to conventional connections can be done quickly and easily.
- Easy to use
- Reliable connecton
- Leak free every time
- Removes contamination caused by Vespel™ or graphite ferrules
- No need to re-tighten ferrules after temperature cycling
- Reduce air background
- NO TOOLS (FingerTite)
Specifications
Thermo INJ/ISQ Starter Kit | |
— | |
0.53 mm I.D. Columns |