missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Shimadzu Splitless Straight-Through Liner

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan™ 0920861

 View more versions of this product

Product Code. 12914021

  • £66.10 / Pack of 5
Estimated Shipment: 17-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with Shimazdu instruments
Specifications

Specifications

5/Pk.
Spitless Straight-through Liner
5 mm
95 mm
2.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers