missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ SilTite™ GC Capillary Column Connectors: μ Unions

SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools

Brand:  Trajan™ 073564

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15268933

  • £144.00 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

  • Low thermal mass
  • Zero Dead Volume
  • Inert
  • No-need to retighten after installation
  • Connect Pre-column or GC x GC
Specifications

Specifications

Connector
0.32 mm and 0.53 mm
2 unions, 5 double taper ferrules and tools
GC Capillary Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers