Learn More
SGE™ ConnecTite™ Liners for Thermo Scientific™ GC
Designed for use with Thermo Scientific TRACE™ and FOCUS™ GCs
Brand: SGE 092334
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Range covers all applications and all popular instrument makes/ models
- Range covers vaporizing and cool on-column injection techniques
- Inlet liners ensure complete transfer of sample from syringe to the column without loss
- 8mm OD, 5mm ID; 105mm length
Specifications
Connectite Standard Liner | |
10.5 cm | |
0.8 cm |
5/Pk. | |
0.5 cm | |
Thermo Scientific GC Trace and Focus |