missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPC Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10774-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody (Product No. 10774-1-AP) is a rabbit polyclonal antibody raised with the full-length SFTPC of human origin.
This gene encodes the pulmonary-associated surfactant protein C (SPC), an extremely hydrophobic surfactant protein essential for lung function and homeostasis after birth. Pulmonary surfactant is a surface-active lipoprotein complex composed of 90% lipids and 10% proteins which include plasma proteins and apolipoproteins SPA, SPB, SPC and SPD. The surfactant is secreted by the alveolar cells of the lung and maintains the stability of pulmonary tissue by reducing the surface tension of fluids that coat the lung. Multiple mutations in this gene have been identified, which cause pulmonary surfactant metabolism dysfunction type 2, also called pulmonary alveolar proteinosis due to surfactant protein C deficiency, and are associated with interstitial lung disease in older infants, children, and adults. Alternatively spliced transcript variants encoding different protein isoforms have been identified.Specifications
| SFTPC | |
| Polyclonal | |
| Unconjugated | |
| SFTPC | |
| PSP C, SFTP2, SFTPC, SMDP2, SP C, SP5, surfactant protein C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 50683, 6440 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11685, P11686 | |
| SFTPC | |
| SFTPC Fusion Protein Ag1159 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction