missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | SFRS8 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SFRS8 Polyclonal specifically detects SFRS8 in Human samples. It is validated for Western Blot.Specifications
| SFRS8 | |
| Polyclonal | |
| Rabbit | |
| B2RN45 | |
| 6433 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Drosophila homolog), Drosophila), SFRS8, Splicing factor, arginine/serine-rich 8, splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot, splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, splicing factor, suppressor of white-apricot homolog, splicing factor, suppressor of white-apricot homolog (Drosophila), Suppressor of white apricot protein homolog, SWAPMGC167082 | |
| SFSWAP | |
| Primary |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit