missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57466
This item is not returnable.
View return policy
Description
SFRS8 Polyclonal specifically detects SFRS8 in Human samples. It is validated for Western Blot.
Specifications
| SFRS8 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Drosophila homolog), Drosophila), SFRS8, Splicing factor, arginine/serine-rich 8, splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot, splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, splicing factor, suppressor of white-apricot homolog, splicing factor, suppressor of white-apricot homolog (Drosophila), Suppressor of white apricot protein homolog, SWAPMGC167082 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6433 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| B2RN45 | |
| SFSWAP | |
| Synthetic peptides corresponding to SFRS8(splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, Drosophila)) The peptide sequence was selected from the N terminal of SFRS8. Peptide sequence LVFGYACKLFRDDERALAQEQGQHLIPWMGDHKILIDRYDGRGHLHDLSE The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction