Learn More
Vapourtec Ltd. SF-10 Solvent/Reagent Valve Kit
Click to view available options
For Use With:
SF-10 Reagent Pump
Description
- A Waste/collect selection valve ca be fitted to the SF-10 pump to allow automated switching of the output stream to a collection vessel or another instrument
Specifications
Specifications
| For Use With | SF-10 Reagent Pump |
| Type | Solvent/Reagent Valve Kit |