Learn More
Mettler Toledo™ SevenGo Duo pro™ SG68 Field Kit
Stores up to 500 GLP records for mobile measurement of pH, mV, ion concentration and dissolved oxygen. Mettler Toledo™ SevenGo Duo pro™ SG68 Field Kit stores readings with the time/date, sensor ID and SN, and user and sample IDs. The instrument provides contact-free IR communication.
Brand: Mettler Toledo™ 51302612
Code : 59
Additional Details : Weight : 0.40000kg
Description
The SG68 SevenGo Duo pro™ pH/ion/oxygen meter meets the highest requirements for measurements in dual-channel mode. It allows simultaneous determination and display of two electrochemical parameters with the highest possible accuracy. This instrument has 5-point pH calibration with a choice of 7 predefined and 1 user-defined buffer sets, and has an integrated barometer and polarographic oxygen sensor for measuring dissolved oxygen.
The SG68 SevenGo Duo pro™ features extra security functions, such as PIN-protected device logon and PIN-protected system setting. The Intelligent Sensor Management (ISM™) safety feature automatically detects the connected sensor and uses the most up-to-date calibration data stored on the sensor chip.
- Freedom of communication: Infrared technology enables wireless transmission of data to a laptop, printer and PC
- GLP excellence: All relevant information is stored for each measurement and can be printed
- PC Software – LabX™ direct pH: User-friendly PC software archives results, with easy transfer to an open application such as MS Excel™
- Durable uGo carrying case: Securely stores all hardware in a hermetically-sealed interior
- Work in the field or lab: Single keystroke functionality for routine mode or an expert mode that uses the entire range of functions
- At a glance: Large display with a bright backlight and a 10-language menu to show all items
- Ergonomic masterpiece: Fits neatly into either hand with a rotatable electrode clip that can be attached on either side
- Watertight and dustproof: The entire system follows the IP (Ingress Protection) 67 code
- Complete field kit: Includes instrument, InLab Expert Pro sensors with ISM™, 1.8m or 5m fixed cable, electrode clip, uGo case, and calibration bottles and standards
Specifications
IR to printer or PC via RS232 or USB | |
±0.5% | |
±0.002 | |
1.5 V | |
5 | |
Dual-channel | |
IP 67 | |
0.001 | |
Suited for mobile measurements both in the field and in the production environment | |
pH / DO sensor inputs: BNC, RCA/cinch (both IP67) | |
500 measurements | |
4 AA batteries or 1.3V NiMH accumulators | |
1 | |
0.000 to 999.9% 0.000 to 9999 ppm 1.00E-9 to 9.99E+9 |
|
-1999 to 1999 mV | |
0.1 | |
0.0 to 600% | |
±0.2°C | |
0.1°C | |
368 g (Without batteries) |
±1 | |
±0.1 | |
±0.5% max 0.03 | |
5 | |
SevenGo Duo pH/Ion/DO meter SG68-FK2-Kit | |
LCD | |
-2.000 to 19.999 | |
-5°C to +130°C (ATC), -30°C to +130°C (MTC) | |
InLab Expert Pro-ISM and InLab605-ISM (both IP67, 1.8m cable) and uGo | |
Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese | |
SG68-FK2 | |
500 to 100 mbar | |
Field Kit | |
0.00 to 99.00 mg/L | |
5–85% (non-cond.) | |
0.01 | |
0.1 to 1 | |
0°C to 40°C | |
220 x 90 x 45mm |