missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ SevenGo Duo pro™ SG68 Field Kit

Product Code. 10004414 Shop All Mettler Toledo Products
Change view
Click to view available options
Product Type:
Meter with Field Kit, 2m Cable
Meter with Field Kit, 5m Cable
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
10004414 Meter with Field Kit, 5m Cable Each
10799175 Meter with Field Kit, 2m Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10004414 Supplier Mettler Toledo™ Supplier No. 51302613
Add to basket
Add to basket
This item is not returnable. View return policy

Stores up to 500 GLP records for mobile measurement of pH, mV, ion concentration and dissolved oxygen. Mettler Toledo™ SevenGo Duo pro™ SG68 Field Kit stores readings with the time/date, sensor ID and SN, and user and sample IDs. The instrument provides contact-free IR communication.

The SG68 SevenGo Duo pro™ pH/ion/oxygen meter meets the highest requirements for measurements in dual-channel mode. It allows simultaneous determination and display of two electrochemical parameters with the highest possible accuracy. This instrument has 5-point pH calibration with a choice of 7 predefined and 1 user-defined buffer sets, and has an integrated barometer and polarographic oxygen sensor for measuring dissolved oxygen.

The SG68 SevenGo Duo pro™ features extra security functions, such as PIN-protected device logon and PIN-protected system setting. The Intelligent Sensor Management (ISM™) safety feature automatically detects the connected sensor and uses the most up-to-date calibration data stored on the sensor chip. 

  • Freedom of communication: Infrared technology enables wireless transmission of data to a laptop, printer and PC 
  • GLP excellence: All relevant information is stored for each measurement and can be printed
  • PC Software – LabX™ direct pH: User-friendly PC software archives results, with easy transfer to an open application such as MS Excel™
  • Durable uGo carrying case: Securely stores all hardware in a hermetically-sealed interior
  • Work in the field or lab: Single keystroke functionality for routine mode or an expert mode that uses the entire range of functions
  • At a glance: Large display with a bright backlight and a 10-language menu to show all items
  • Ergonomic masterpiece: Fits neatly into either hand with a rotatable electrode clip that can be attached on either side
  • Watertight and dustproof: The entire system follows the IP (Ingress Protection) 67 code
  • Complete field kit: Includes instrument, InLab Expert Pro sensors with ISM™, 1.8m or 5m fixed cable, electrode clip, uGo case, and calibration bottles and standards
TRUSTED_SUSTAINABILITY

Specifications

Outputs IR to printer or PC via RS232 or USB
Accuracy (Barometer) ±1
Accuracy (ISE) ±0.5%
Accuracy (mV) ±0.1
Accuracy (pH) ±0.002
Accuracy (Polarographic DO) ±0.5% max 0.03
Battery Type 1.5 V
Calibration Points 5
Calibration Points (pH) 5
Description SevenGo Duo pH/Ion/DO meter SG68-FK5-Kit
Channels 2
Display Type LCD
IP Rating IP67
Range (pH) -2.000 to 19.999
Resolution (pH) 0.001
Temperature Range (Metric) -5°C to +130°C (ATC), -30°C to +130°C (MTC)
For Use With (Application) Suited for mobile measurements both in the field and in the production environment
Includes InLab Expert Pro-ISM and InLab605-ISM (both IP67, 5m cable) and uGo
Inputs pH / DO sensor inputs: BNC, RCA/cinch (both IP67)
Languages Chinese, Chinese (TW), Croatian, Czech, Danish, Dutch, English (GB), English (US), French, German, Hungarian, Indonesian, Italian, Japanese, Korean, Norwegian, Polish, Portuguese (BR), Russian, Slovak, Slovenian, Spanish, Swedish, Thai, Turkish, Vietnamese
Memory 500 measurements
Model SG68-FK5
Power Supply 4 AA batteries or 1.3V NiMH accumulators
Pressure Range 500 to 100 mbar
Pressure Resolution 1
Product Type Meter with Field Kit, 5m Cable
Range (ISE) 0.000 to 999.9%
0.000 to 9999 ppm
1.00E-9 to 9.99E+9
Range (Polarographic DO) 0.00 to 99.00 mg/L
Range (Relative mV) -1999 to 1999 mV
Relative Humidity Range 5–85% (non-cond.)
Resolution (mV) 0.1
Resolution (Polarographic DO) 0.01
Saturation Range 0.0 to 600%
Saturation Resolution 0.1 to 1
Temperature Accuracy ±0.2°C
Temperature (Metric) Operating 0°C to 40°C
Temperature Resolution 0.1°C
Dimensions (L x W x H) 220 x 90 x 45mm
Weight (Metric) 368 g (Without batteries)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.