Learn More
Thermo Scientific™ Settling Plate, Sterile, Tryptic Soy Agar (Irradiated)
Product Code. 17415539
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
150mm Plate - Double + Bag
Monoplate - Double + Bag
Unit Size:
Pack of 10
Pack of 20
Product Code. 17415539
Brand: Thermo Scientific™ R111874
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Sterile Tryptic Soy Agar (Irradiated).
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Sterile Tryptic Soy Agar (Irradiated). It is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. It is also recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.
Tryptic Soy Agar is prepared following the standard formula for Soybean-Casein Digest Agar Medium in the United States of Pharmacopeia (USP)5. Tryptic Soy Agar many be supplemented with blood to facilitate the growth of more fastidious bacteria or antimicrobial agents to permit the selection of various microbial groups from mixed flora.
  • Recommended for use in Official Methods of Analysis of AOAC International, IDF, and USDA3,4
  • Ready-to-use – Convenience of prepared medium
This medium contains casein and soy peptones that provide nitrogen, amino acids, and peptides necessary for bacterial growth. Dextrose is a ready source of energy. Sodium chloride supplies essential electrolytes and maintains osmotic equilibrium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Settling Plate Sterile Tryptic Soy Agar
Format 150mm Plate - Double + Bag
Product Type Agar
Quantity 20/Pk.
Not intended for IVD use.