missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-80046
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
SETD4 Polyclonal specifically detects SETD4 in Human samples. It is validated for Western Blot.
Specifica
| SETD4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C21orf18, C21orf27, chromosome 21 open reading frame 18, chromosome 21 open reading frame 27, SET domain containing 4, SET domain-containing protein 4 | |
| Rabbit | |
| 50 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_059134 | |
| SETD4 | |
| Synthetic peptide directed towards the C terminal of human C21ORF18. Peptide sequence LTALKLLCLEAEKFTCWKKVLLGEVISDTNEKTSLDIAQKICYYFIEETN. | |
| Affinity purified | |
| RUO | |
| 54093 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto