missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SESN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£200.00 - £558.00
Specifications
| Antigen | SESN3 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Target Species | Human, Mouse, Rat |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18063486
|
Novus Biologicals
NBP1-44985-50UG |
50 μg |
£342.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18250893
|
Novus Biologicals
NBP1-44985-100UG |
100 μg |
£558.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18215084
|
Novus Biologicals
NBP1-44985-0.025MG |
0.025 mg |
£200.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SESN3 Polyclonal specifically detects SESN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| SESN3 | |
| Unconjugated | |
| Human, Mouse, Rat | |
| SEST3MGC29667, sestrin 3, sestrin-3 | |
| SESN3 | |
| IgG |
| Polyclonal | |
| Rabbit | |
| P58005 | |
| 143686 | |
| Synthetic peptides corresponding to unique epitope on SESN3. The SESN3 peptide was selected from N-terminal, amino acids 1-50. This peptide was covalently modified post-synthetically to achieve desired antigenicity. | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title