missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SESN3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-44985-100UG
This item is not returnable.
View return policy
Description
SESN3 Polyclonal specifically detects SESN3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| SESN3 | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% Sodium Azide | |
| SEST3MGC29667, sestrin 3, sestrin-3 | |
| Rabbit | |
| 57 kDa | |
| 100 μg | |
| Cell Cycle and Replication | |
| 143686 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Dot Blot | |
| 0.5 mg/mL | |
| Western Blot >1:500, ELISA < 1:10000, Dot Blot 1:10000 | |
| P58005 | |
| SESN3 | |
| Synthetic peptides corresponding to unique epitope on SESN3. The SESN3 peptide was selected from N-terminal, amino acids 1-50. This peptide was covalently modified post-synthetically to achieve desired antigenicity. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction