Learn More
VACUUBRAND™ Service Kits for Oil-Free Diaphragm Vacuum Pumps
Product Code. 10486153 Shop All Vacuubrand Products
Click to view available options
For Use With:
MD 4 NT/VARIO, MV 2 NT/VARIO
MD1C, PC3001 Diaphragm Pump
ME 8 NT
ME1, ME1C Diaphragm Pump
ME2C NT Diaphragm Pump
MZ 2 NT, ME 4 NT + VARIO
MZ 4C-MD 4C
MZ2D NT VARIO
Unit Size:
1 set
Each
Product Code. 10486153
Brand: VACUUBRAND™ 20696861
Only null left
Add to basket
Add to basket
Original VACUUBRAND replacement parts
  • Includes appropriate diaphragms, valves, and sealing rings, if appropriate
  • Does not include spanner wrench
TRUSTED_SUSTAINABILITY
Specifications
For Use With MD 4 NT/VARIO, MV 2 NT/VARIO
Type Service Kit