missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SerpinB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | SerpinB2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SerpinB2 Polyclonal specifically detects SerpinB2 in Human samples. It is validated for Western Blot.Specifications
| SerpinB2 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer | |
| HsT1201, Monocyte Arg-serpin, PAI, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2 | |
| SERPINB2 | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| P05120 | |
| 5055 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title