missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SerpinB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59086
This item is not returnable.
View return policy
Description
SerpinB2 Polyclonal specifically detects SerpinB2 in Human samples. It is validated for Western Blot.
Specifications
| SerpinB2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| HsT1201, Monocyte Arg-serpin, PAI, PAI2Urokinase inhibitor, Placental plasminogen activator inhibitor, PLANH2PAI-2, plasminogen activator inhibitor 2, plasminogen activator inhibitor, type II (arginine-serpin), serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2, Serpin B2, serpin peptidase inhibitor, clade B (ovalbumin), member 2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Mouse: 100%; Rabbit: 100%; Guinea pig: 92%; Equine: 92%; Pig: 92%; Bovine: 85%; Sheep: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P05120 | |
| SERPINB2 | |
| Synthetic peptides corresponding to SERPINB2(serpin peptidase inhibitor, clade B (ovalbumin), member 2) The peptide sequence was selected from the middle region of SERPINB2. Peptide sequence GKIPNLLPEGSVDGDTRMVLVNAVYFKGKWKTPFEKKLNGLYPFRVNSAQ The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Apoptosis, Cancer | |
| 5055 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction