missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A1/alpha 1-Antitrypsin Rabbit anti-Human, Clone: 142, Novus Biologicals™
Rabbit Monoclonal Antibody
£307.00 - £448.00
Specifications
| Antigen | Serpin A1/alpha 1-Antitrypsin |
|---|---|
| Clone | 142 |
| Dilution | Sandwich ELISA Capture 1:250-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18260447
|
Novus Biologicals
NBP2-89402-50UL |
50 μg |
£307.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18254536
|
Novus Biologicals
NBP2-89402-100UL |
100 μg |
£448.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for Sandwich ELISA Capture.Specifications
| Serpin A1/alpha 1-Antitrypsin | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Recombinant Human Serpin A1/alpha 1-Antitrypsin Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 142 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 5265 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title