missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serpin A1/alpha 1-Antitrypsin Rabbit anti-Human, Clone: 142, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89402-100UL
This item is not returnable.
View return policy
Description
Serpin A1/alpha 1-Antitrypsin Monoclonal specifically detects Serpin A1/alpha 1-Antitrypsin in Human samples. It is validated for Sandwich ELISA Capture.
Specifications
| Serpin A1/alpha 1-Antitrypsin | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 142 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| A1A, AATMGC23330, Alpha-1 protease inhibitor, Alpha-1-antiproteinase, alpha-1-antitrypsin, antitrypsin), member 1, member 1, PIMGC9222, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serine (or cysteine) proteinase inhibitor, clade A, member 1, Serpin A1, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin) | |
| Recombinant Human Serpin A1/alpha 1-Antitrypsin Protein | |
| 100 μg | |
| Stem Cell Markers | |
| 5265 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction