missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serotonin Mouse anti-Human, Mouse, Porcine, Chicken, Macaque, Clone: 5HT-H209, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 13 publications
£247.89 - £1030.00
Specifications
| Antigen | Serotonin |
|---|---|
| Clone | 5HT-H209 |
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:50-1:100 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18241923
|
Novus Biologicals
NB120-16007-500UL |
500 μL |
£1030.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18088413
|
Novus Biologicals
NB120-16007-0.1ML |
0.1 mL | |||||||
Description
Serotonin Monoclonal specifically detects Serotonin in Human, Mouse, Porcine, Chicken, Rhesus Macaque samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Serotonin | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:50-1:100 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| 5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin). | |
| Primary | |
| Store at 4°C. Do not freeze. |
| 5HT-H209 | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Cell culture supernatant dialysed against 0.05 mol/L Tris/HCl (pH 7.2) | |
| IgG1 κ | |
| Tissue culture supernatant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title