missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serotonin Mouse anti-Human, Mouse, Porcine, Chicken, Macaque, Clone: 5HT-H209, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 13 publications
Brand: Novus Biologicals NB120-16007-500UL
This item is not returnable.
View return policy
Description
Serotonin Monoclonal specifically detects Serotonin in Human, Mouse, Porcine, Chicken, Rhesus Macaque samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Serotonin | |
| Monoclonal | |
| Unconjugated | |
| Cell culture supernatant dialysed against 0.05 mol/L Tris/HCl (pH 7.2) | |
| 5-Hydroxytryptamine hydrochloride (3-(2-aminoethyl)-5-hydroxyindole;serotonin). | |
| 500 μL | |
| Primary | |
| Store at 4°C. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 5HT-H209 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:50-1:100 | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Human, Mouse, Pig, Chicken, Macaque | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction