missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Septum for 8-425 screw thread cap

Product Code. 11850772 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Beige, Red
Beige, Red-Orange
Black
Blue Transparent, White
Blue, White
Cream, Red
Dark Blue, White
Gray, Red
Red, White
Red-Orange, Transparent
White
Material:
Butyl, PTFE
Natural Rubber, TEF
PTFE
PTFE Virginal
PTFE, Rubber
PTFE, Silicone
Viton 1A
Thickness (Metric):
0.25 mm
0.9 mm
1 mm
1.3 mm
1.5 mm
Unit Size:
Pack of 100
Pack of 1000
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material Thickness (Metric) unitSize
11850772 Blue Transparent, White PTFE, Silicone 1.3 mm Pack of 1000
11787557 Red-Orange, Transparent Natural Rubber, TEF 1.3 mm Pack of 1000
11841653 Blue, White PTFE, Silicone 0.9 mm Pack of 1000
11872663 Red, White PTFE, Silicone 1.3 mm Pack of 1000
11821663 White PTFE Virginal 0.25 mm Pack of 1000
11828961 Red, White PTFE, Silicone 1 mm Pack of 1000
11830772 Cream, Red PTFE, Silicone 1.5 mm Pack of 1000
16160961 Beige, Red-Orange PTFE 1 mm Pack of 1000
12950841 Beige, Red-Orange PTFE 1 mm Pack of 100
11858961 Black Viton 1A 1 mm Pack of 1000
11838961 Black Viton 1A 1.5 mm Pack of 1000
11840772 Dark Blue, White PTFE, Silicone 1.3 mm Pack of 1000
11868961 Gray, Red Butyl, PTFE 1.3 mm Pack of 1000
10258004 Beige, Red PTFE, Rubber 1 mm Pack of 1000
11848961 Red-Orange, Transparent Natural Rubber, TEF 1 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 11850772

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

  • 8mm Septa
TRUSTED_SUSTAINABILITY

Specifications

Description Septa
Color Blue Transparent, White
Material PTFE, Silicone
For Use With (Application) 8mm
Quantity 1000
Thickness (Metric) 1.3 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.