missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Septum for 13mm Crimp Cap

Product Code. 11814960 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Blue, Gray
Gray, Red
Red-Orange, Transparent
Material:
Butyl, PTFE
Natural Rubber, TEF
Thickness (Metric):
1.3 mm
1.6 mm
2 mm
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material Thickness (Metric) Unit Size
11814960 Blue, Gray Butyl, PTFE 2 mm Pack of 1000
11824960 Red-Orange, Transparent Natural Rubber, TEF 1.3 mm Pack of 1000
11834960 Gray, Red Butyl, PTFE 1.6 mm Pack of 1000
11844960 Gray, Red Butyl, PTFE 2 mm Pack of 1000
11854960 Gray, Red Butyl, PTFE 1.3 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11814960

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

  • 13mm
  • Carrier materials have different physical and chemical properties, temperature resistance, resealability, cleanliness, hardness, thickness
TRUSTED_SUSTAINABILITY

Specifications

Description Septa
Color Blue, Gray
Diameter (Metric) Outer 13 mm
Material Butyl, PTFE
Quantity 1000
Thickness (Metric) 2 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.