missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Septin-8 Recombinant Rabbit Monoclonal Antibody (23GB4905), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552738
This item is not returnable.
View return policy
Description
SEPT8 is a member of the highly conserved septin family. Septins are 40- to 60-kD GTPases that assemble as filamentous scaffolds. They are involved in the organization of submembranous structures, in neuronal polarity, and in vesicle trafficking (Blaser et al., 2003 [PubMed 12909369]).
Specifications
| Septin-8 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SEPTIN8 | |
| 2-Sep; AW046166; EMBL:AAI58797.1, ECO:0000312; KIAA0202; mKIAA0202; RGD:1308449}; SEP2; Sepl; Sept8; sept8 {ECO:0000312; septin 8; Septin8; septin-8; septin-8 {ECO:0000250; UniProtKB:Q8CHH9} | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20362, 23176, 303135 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4905 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B0BNF1, Q8CHH9, Q92599 | |
| SEPTIN8 | |
| A synthesized peptide derived from human Septin 8 (50-85AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction