missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Septin-8 Recombinant Rabbit Monoclonal Antibody (23GB4905), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Septin-8 |
|---|---|
| Clone | 23GB4905 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088493
![]() |
Invitrogen™
MA552738 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089438
|
Invitrogen™
MA552739 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Septin-8 Recombinant Monoclonal antibody specifically detects Septin-8 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Septin-8 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| B0BNF1, Q8CHH9, Q92599 | |
| 20362, 23176, 303135 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4905 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2-Sep; AW046166; EMBL:AAI58797.1, ECO:0000312; KIAA0202; mKIAA0202; RGD:1308449}; SEP2; Sepl; Sept8; sept8 {ECO:0000312; septin 8; Septin8; septin-8; septin-8 {ECO:0000250; UniProtKB:Q8CHH9} | |
| A synthesized peptide derived from human Septin 8 (50-85AA). | |
| Primary | |
| -20°C | |
| SEPTIN8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title