missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52840
This item is not returnable.
View return policy
Description
Septin-6 Polyclonal specifically detects Septin-6 in Human samples. It is validated for Western Blot.
Specifications
| Septin-6 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| 43349 | |
| Synthetic peptides corresponding to SEPT6(septin 6) The peptide sequence was selected from the N terminal of 40427. Peptide sequence TGLGKSTLMDTLFNTKFEGEPATHTQPGVQLQSNTYDLQESNVRLKLTIV. | |
| 100 μL | |
| Cell Cycle and Replication | |
| 23157 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| KIAA0128SEP2septin 2, MGC16619, MGC20339, SEPT2, septin 6, septin-6 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Mouse: 100%; Human: 100%; Bovine: 100%; Canine: 100%; Rat: 92%; Zebra finch: 91%; Sumatran orangutan: 83%; Crab-eating macaque: 83%; Green puffer: 78%; Chicken: 78%; Zebrafish: 78%;. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction