missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NBP1-90094
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Septin-4 Polyclonal specifically detects Septin-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| Septin-4 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Apoptosis-related protein in the TGF-beta signaling pathway, ARTSPeanut-like protein 2, BRADEION, Bradeion beta, Brain protein H5, CE5B3, Cell division control-related protein 2, Cerebral protein 7, H5, hCDCREL-2SEP4, hucep-7, MART, peanut-like 2, peanut-like 2 (Drosophila), PNUTL2CE5B3 beta, septin 4, septin-4, septin-M | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| SEPTIN4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:YDDDLEFRPPSRPQSSDNQQYFCAPAPLSPSARPRSPWGKLDPYDSSEDDKEYVGFATLPNQVHRKS | |
| 0.1 mL | |
| Apoptosis | |
| 5414 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction