Learn More
SEPT9 Rabbit, anti-Human, Clone: 20000000, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82844-2-RR-100UL
Description
The SEPT9 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human SEPT9. This antibody recognizes human, mouse antigen. The SEPT9 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| SEPT9 | |
| 20000000 | |
| AF17q25, FLJ75490, KIAA0991, MLL septin like fusion protein, MSF, MSF1, NAPB, Ov/Br septin, Ovarian/Breast septin, PNUTL4, SEPT9, SeptD1, septin 9, Septin D1, SINT1 | |
| 100 μL | |
| 10801 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunoprecipitation, ELISA | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |