missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT9 Rabbit, anti-Human, Clone: 20000000, Proteintech
Rabbit Recombinant Monoclonal Antibody
£135.00 - £360.00
Specifications
| Antigen | SEPT9 |
|---|---|
| Clone | 20000000 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunoprecipitation, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18046375
|
Proteintech
82844-2-RR-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18060546
|
Proteintech
82844-2-RR-100UL |
100 μL |
£360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SEPT9 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human SEPT9. This antibody recognizes human, mouse antigen. The SEPT9 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| SEPT9 | |
| 500 μg/mL | |
| Liquid | |
| Human, Mouse | |
| AF17q25, FLJ75490, KIAA0991, MLL septin like fusion protein, MSF, MSF1, NAPB, Ov/Br septin, Ovarian/Breast septin, PNUTL4, SEPT9, SeptD1, septin 9, Septin D1, SINT1 | |
| Primary |
| 20000000 | |
| Western Blot, Immunoprecipitation, ELISA | |
| Rabbit | |
| 10801 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title