missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sephaorose™ Fast Flow Anion Exchange Media: ANX

Used for preparative protein separations in both research and industrial applications

Brand:  Cytiva 17-1287-01

Code : GE

Additional Details : CAS Number : 64-17-5 Weight : 0.78800kg


 View more versions of this product

Product Code. 10231180

  • £671.00 / 500mL
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

  • Hydrophilic nature of base matrix ensures low levels of non-specific binding leading to low levels of host cell-derived impurities in the elution pool
  • Matrix is composed of crosslinked 4% agarose beads
  • Part of Sepharose Fast Flow ion exchange platform
  • High chemical stability allows well proven cleaning-in-place (CIP) and sanitization protocols
  • Available in range of pre-packed process development tools

  • Ligand: Diethylaminopropyl
  • Ion exchanger type: Weak anion exchanger
  • Ionic capacity: 0.13 to 0.17mmol Cl-/mL medium
  • Binding capacity: 5mg thyroglobulin/mL medium
  • Pressure/Flow specification: min. 200cm/hr., 100kPa, XK 50/60 column, bed height 25cm

Capture and purification of proteins.

Specifications

Specifications

500 mL
Weak
90 μm
3 to 13 pH
For Protein Separations in Research/Industrial Applications
ANX Sepharose 4 Fast Flow
Crosslinked 4% agarose beads w/diethylaminopropyl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ANX Sepharose

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers