Learn More
Eppendorf™ Sensor Unit 384-well
Product Code. 15768409 Shop All Eppendorf Products
Change view
Click to view available options
Description:
ISO 17025
ISO 9001
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15768409 ISO 17025 Each
15758409 ISO 9001 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15768409 Supplier Eppendorf™ Supplier No. 6328008503
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for use with an X50 series PCR Mastercycler 384-well thermoblock.
  • Sensor unit for 384-well thermoblock
  • Suitable for use with an X50 series Mastercycler
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) X50 series PCR Mastercycler 384-well Thermoblock
Description ISO 17025