Learn More
Eppendorf™ Sensor Unit 384-well
Shop All Eppendorf Products
Click to view available options
Description:
ISO 17025
ISO 9001
Unit Size:
Each
Description
- Sensor unit for 384-well thermoblock
- Suitable for use with an X50 series Mastercycler
Specifications
Specifications
| For Use With (Equipment) | X50 series PCR Mastercycler 384-well Thermoblock |
| Description | ISO 9001 |