Learn More
Eppendorf™ Sensor Plate Flat
Product Code. 15788409 Shop All Eppendorf Products
Change view
Click to view available options
Description:
ISO 17025
ISO 9001
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15788409 ISO 17025 Each
15778409 ISO 9001 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15788409 Supplier Eppendorf™ Supplier No. 6328009500
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable plate for use with an X50 series PCR Mastercycler flat thermoblock.
  • Sensor plate for a flat thermoblock
  • Suitable for use with an X50 series Mastercycler
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) X50 series PCR Mastercycler Flat Thermoblock
Description ISO 17025