missing translation for 'onlineSavingsMsg'
Learn More

Semenogelin II Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £366.00

Specifications

Antigen Semenogelin II
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18605421
View Documents
Novus Biologicals
NBP2-93405-0.02ml
0.02 mL
£151.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18673871
View Documents
Novus Biologicals
NBP2-93405-0.1ml
0.1 mL
£366.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Semenogelin II Polyclonal antibody specifically detects Semenogelin II in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

Semenogelin II
Western Blot
Unconjugated
Rabbit
Cell Biology, Cytoskeleton Markers, Signal Transduction
PBS (pH 7.3), 50% glycerol
6407
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human, Mouse
semenogelin IISGIISemenogelin 2, semenogelin-2
Recombinant fusion protein containing a sequence corresponding to amino acids 383-582 of human SEMG2 (NP_002999.1). HGKSQNQVRIPSQAQEYGHKENKISYQSSSTEERRLNSGEKDVQKGVSKGSISIQTEEKIHGKSQNQVTIPSQDQEHGHKENKMSYQSSSTEERRLNYGGKSTQKDVSQSSISFQIEKLVEGKSQIQTPNPNQDQWSGQNAKGKSGQSADSKQDLLSHEQKGRYKQESSESHNIVITEHEVAQDDHLTQQYNEDRNPIST
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.