missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4D/CD100 Antibody (A8), Novus Biologicals™
Mouse Monoclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | Semaphorin 4D/CD100 |
|---|---|
| Clone | A8 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Flow (Cell Surface) |
| Applications | Flow Cytometry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18122185
|
Novus Biologicals
NBP2-25198SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18103193
|
Novus Biologicals
NBP2-25198 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Flow (Cell Surface).Specifications
| Semaphorin 4D/CD100 | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Immunology | |
| NP_006369 | |
| 10507 | |
| PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| A8 | |
| Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Flow (Cell Surface) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
| SEMA4D | |
| IgG1 κ | |
| Protein G purified | |
| This product reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title