missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4D/CD100 Antibody (A8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25198SS
This item is not returnable.
View return policy
Description
Semaphorin 4D/CD100 Monoclonal specifically detects Semaphorin 4D/CD100 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Flow (Cell Surface).
Specifications
| Semaphorin 4D/CD100 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Flow (Cell Surface) | |
| A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Human. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunoprecipitation | |
| A8 | |
| Unconjugated | |
| NP_006369 | |
| SEMA4D | |
| PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody. | |
| 0.025 mg | |
| Apoptosis, Immunology | |
| 10507 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction