Learn More
Invitrogen™ SEMA4A Monoclonal Antibody (KL-1 (aka SK31)), eBioscience™

Rat Monoclonal Antibody
£239.00
Specifications
| Antigen | SEMA4A |
|---|---|
| Clone | KL-1 (aka SK31) |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C |
| Applications | Western Blot |
Description
Description: The KL-1 (aka SK31) antibody reacts with mouse Sema4A, a class IV semaphorin originally identified as SemB. Sema4A is structurally similar to Sema4D/CD100. Sema4A is reported to be expressed by dendritic cells, B cells and activated T cells. Although published data suggested that Sema4A binds to Tim-2, a T cell surface receptor, it remains to be definitively shown that these two molecules have a receptor/ligand relation. It is suggested that Sema4a can provide costimulatory signals for T cells. This KL-1 antibody works in immunoblotting but requires non-reducing conditions. Applications Reported: This KL-1 (aka SK31) antibody has been reported for use in immunoblotting (WB). Applications Tested: The KL-1 (aka SK31) antibody has been tested by immunoblot analysis of mouse Sema4A transfected cells under nonreducing conditions. This can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
SEMA4A is a member of the semaphorin family of soluble and transmembrane proteins. Semaphorins are involved in guidance of axonal migration during neuronal development and in immune responses.Specifications
| SEMA4A | |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Q62178 | |
| 20351 | |
| Primary | |
| Affinity chromatography |
| KL-1 (aka SK31) | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG2a | |
| Mouse | |
| SEMA4A | |
| AI132332; CORD10; RP35; Sema B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4A; Sema4a; Semab; semaphorin 4A; semaphorin B; semaphorin-4A; Semaphorin-B; SemB; UNQ783/PRO1317 | |
| SEMA4A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.