missing translation for 'onlineSavingsMsg'
Learn More

SEL1L Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£170.00 - £395.00

Specifications

Antigen SEL1L
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18654261
View Documents
Novus Biologicals
NBP2-93746-0.02ml
0.02 mL
£170.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18607691
View Documents
Novus Biologicals
NBP2-93746-0.1ml
0.1 mL
£395.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SEL1L Polyclonal antibody specifically detects SEL1L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

SEL1L
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Cancer, Immunology, Signal Transduction
PBS (pH 7.3), 50% glycerol
6400
IgG
Affinity purified
Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
IBD2, PRO1063, protein sel-1 homolog 1, sel-1 (suppressor of lin-12, C.elegans)-like, sel-1 suppressor of lin-12-like (C. elegans), sel-1 suppressor of lin-12-like 1, Sel-1L, SEL1L1, SEL1-LIKE, Suppressor of lin 12 (sel-1), C. elegans, homolog of, Suppressor of lin-12-like protein 1, TSA305
Recombinant fusion protein containing a sequence corresponding to amino acids 22-190 of human SEL1L (NP_005056.3). SSDEEGSQDESLDSKTTLTSDESVKDHTTAGRVVAGQIFLDSEESELESSIQEEEDSLKSQEGESVTEDISFLESPNPENKDYEEPKKVRKPALTAIEGTAHGEPCHFPFLFLDKEYDECTSDGREDGRLWCATTYDYKADEKWGFCETEEEAAKRRQMQEAEMMYQTG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.