missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEL1L Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93746-0.02ml
This item is not returnable.
View return policy
Description
SEL1L Polyclonal antibody specifically detects SEL1L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SEL1L | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| IBD2, PRO1063, protein sel-1 homolog 1, sel-1 (suppressor of lin-12, C.elegans)-like, sel-1 suppressor of lin-12-like (C. elegans), sel-1 suppressor of lin-12-like 1, Sel-1L, SEL1L1, SEL1-LIKE, Suppressor of lin 12 (sel-1), C. elegans, homolog of, Suppressor of lin-12-like protein 1, TSA305 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-190 of human SEL1L (NP_005056.3). SSDEEGSQDESLDSKTTLTSDESVKDHTTAGRVVAGQIFLDSEESELESSIQEEEDSLKSQEGESVTEDISFLESPNPENKDYEEPKKVRKPALTAIEGTAHGEPCHFPFLFLDKEYDECTSDGREDGRLWCATTYDYKADEKWGFCETEEEAAKRRQMQEAEMMYQTG | |
| 0.02 mL | |
| Cancer, Immunology, Signal Transduction | |
| 6400 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction