missing translation for 'onlineSavingsMsg'
Learn More

Securin / PTTG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen Securin / PTTG1
Concentration 0.38 mg/mL
Applications Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16898074
View Documents
Proteintech
18040-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16888074
View Documents
Proteintech
18040-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pituitary tumor-transforming gene 1 (PTTG-1) is an oncogene expressed in normal tissues containing proliferating cells and in tumors. PTTG is downstream of the MAP kinase signaling cascade and is phosphorylated at Ser162 by MAPK in vitro. Phosphorylation plays an essential role in PTTG′s transactivation function. The PTTG protein contains a DNA-binding domain, and it activates transcription of the c-Myc oncogene by binding to the promoter to form a complex. Studies have demonstrated that expression of human PTTG correlates with cell proliferation in a cell cycle dependent manner, and the association of human PTTG with Ku, the regulatory subunit of DNA-dependent protein kinase, is disrupted when a DNA double-strand breaks occurs.
Specifications

Specifications

Securin / PTTG1
Immunofluorescence, Immunocytochemistry
Unconjugated
Rabbit
Mouse, Rat, Human
O95997, P97613, Q9CQJ7
30939, 64193, 9232
Securin / PTTG1 Fusion Protein Ag12670
Primary
-20°C
PTTG1
0.38 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
EAP1, Esp1 associated protein, HPTTG, pituitary tumor transforming 1, PTTG, PTTG1, Securin, Tumor transforming protein 1, TUTR1
Pttg1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.