missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Securin / PTTG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18040-1-AP-150UL
This item is not returnable.
View return policy
Description
Pituitary tumor-transforming gene 1 (PTTG-1) is an oncogene expressed in normal tissues containing proliferating cells and in tumors. PTTG is downstream of the MAP kinase signaling cascade and is phosphorylated at Ser162 by MAPK in vitro. Phosphorylation plays an essential role in PTTG′s transactivation function. The PTTG protein contains a DNA-binding domain, and it activates transcription of the c-Myc oncogene by binding to the promoter to form a complex. Studies have demonstrated that expression of human PTTG correlates with cell proliferation in a cell cycle dependent manner, and the association of human PTTG with Ku, the regulatory subunit of DNA-dependent protein kinase, is disrupted when a DNA double-strand breaks occurs.Specifications
| Securin / PTTG1 | |
| Polyclonal | |
| Unconjugated | |
| PTTG1 | |
| EAP1, Esp1 associated protein, HPTTG, pituitary tumor transforming 1, PTTG, PTTG1, Securin, Tumor transforming protein 1, TUTR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 30939, 64193, 9232 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95997, P97613, Q9CQJ7 | |
| Pttg1 | |
| Securin / PTTG1 Fusion Protein Ag12670 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction