missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ SecureFit™ Protective Eyewear
Click to view available options
Lens Tint:
Amber
Clear
Gray
Unit Size:
Each
Pack of 20
Description
- Need to stock multiple frame sizes at work sites eliminated by self-adjusting feature
- Secure, snug fit prevents slippage even during motion
- Anti-fog coating ensures moisture free lenses in hot and humid conditions
- Anti-scratch coating protects lenses from scratches, nicks and scrapes
- Polycarbonate lenses absorb 99.9% UVA and UVB up to 380nm
- 3M Catalog# SF201AF, SF201AS, SF202AF, SF202AS, SF203AF
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1-2015, CSA Z94.3 |
| Lens Coating | Anti-fog |
| Lens Tint | Amber |
| Model | SF203AF |
| No. per Pack | 20/Pk. |
| Temple Style | Proprietary self-adjusting 3M Pressure Diffusion Temple Technology |
| Lens Material | Polycarbonate |
| Frame Type | Frameless |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto