Learn More
3M™ SecureFit™ Safety Glasses, Anti-Fog / Anti-Scratch Coating (K&N)
Product Code. 17267660
Click to view available options
Frame Color:
Black/Yellow
Brown
Gray/Blue
Grey
Red/Gray
Lens Tint:
Amber
Brown
Clear
Gray
Indoor/Outdoor Gray
Orange
Description:
SF401SGAF-BLU-EU
SF401SGAF-BLU-F-EU, Includes removable foam-lined gasket
SF401SGAF-RED-EU
SF402SGAF-BLU-EU
SF403SGAF-YEL-EU
SF405SGAF-BLA-EU
SF406SGAF-BLA-EU
SF407SGAF-BLU-EU
SF601SGAF/FI-EU
Unit Size:
Each
Pack of 20
Product Code. 17267660
Brand: 3M™ 7100148026
Only null left
Add to basket
Add to basket
Wide flexible temple arms, soft, adjustable nose pads and a clear, anti-scratch and superior anti-fog, rimless polycarbonate lens
  • Diffuse pressure over the ear with 3M™ Pressure Diffusion Temple Technology
  • Soft, adjustable nose pads provide added comfort
  • Flat and flexible temples combine easily with earmuffs and provide solid side protection
  • Wraparound design gives the user a wide field of vision
  • 3M™ Scotchgard™ Anti-Fog Coating offers superior anti-fog and anti-scratch properties
  • Excellent protection against Ultraviolet (UV) radiation
  • 19 g lightweight design
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Description SF401SGAF-RED-EU
Lens Tint Clear
Frame Color Red/Gray
Includes  
Quantity 20/Cs.
Lens Material Polycarbonate
Frame Material Polycarbonate