Description
- Sleek and compatible 3M™ SecureFit™ Safety Glasses range with adjustable ratchet temples to adjust the lens for facial features and eyeglass position, designed to help improve the positioning when worn with respirators
- 3M's innovative SecureFit™ Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalization. The temples flex and self-adjust to the size of wearer's head
- 3M™ Scotchgard™ Anti-Fog Coating provides superior anti-fog and anti-scratch properties (available on selected models)
- Protection against UV up to 400 nm
- Wraparound design improves the security of fit and prevents slipping
- Flat and flexible temples combine easily with earmuffs and provide solid side protection
- Easy to clean
- Foam gasket available to help limit eye exposure to debris
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Certifications/Compliance (Europe) | EN 166:2001 |
| Lens Tint | IR 3.0 Gray |
| Product Type | Protective Eyewear |
| Frame Color | Green, Black |
| Model | SF530ASP-GRN-EU |
| Frame Material | Polycarbonate |
| UV Protection | UV400 |